Assay Details
Gene Symbol: mcm4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005655.1 | NP_001005655.1 | 3 - 4 | 338 | 66 | |
XM_012964516.2 | XP_012819970.1 | 3 - 4 | 392 | 66 | ||
GenBank mRNA | BC074670.1 | - | 3 - 4 | 338 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
