Assay Details
| Gene Symbol: rab27b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005681.2 | NP_001005681.1 | 5 - 6 | 539 | 62 | |
| NM_001195546.1 | NP_001182475.1 | 7 - 8 | 806 | 62 | ||
| GenBank mRNA | BC075102.1 | - | 5 - 6 | 539 | 62 | |
| DN070478.1 | - | 6 - 7 | 633 | 62 | ||
| DT402506.1 | - | 1 - 2 | 698 | 62 | ||
More Information
Set Membership: |
|

rab27b
Genomic Map