Assay Details
Gene Symbol: ccni | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005690.1 | NP_001005690.1 | 5 - 6 | 659 | 59 | |
XM_012959253.1 | XP_012814707.1 | 6 - 7 | 731 | 59 | ||
XM_012959260.2 | XP_012814714.1 | 7 - 8 | 1121 | 59 | ||
GenBank mRNA | BC075116.1 | - | 5 - 6 | 659 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
