Assay Details
| Gene Symbol: lypla1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005699.1 | NP_001005699.1 | 1 - 2 | 124 | 54 | |
| GenBank mRNA | BC075270.1 | - | 1 - 2 | 124 | 54 | |
| CR761348.2 | - | 1 - 2 | 116 | 54 | ||
More Information
Set Membership: |
|

lypla1
Genomic Map