Assay Details
| Gene Symbol: hccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005708.2 | NP_001005708.2 | 2 - 3 | 318 | 93 | |
| NM_001247977.1 | NP_001234906.1 | 2 - 3 | 367 | 93 | ||
| NM_001247978.1 | NP_001234907.1 | 2 - 3 | 321 | 93 | ||
| XM_012960825.2 | XP_012816279.1 | 2 - 3 | 308 | 93 | ||
| GenBank mRNA | AL850176.2 | - | 2 - 3 | 321 | 93 | |
| BC075286.1 | - | 2 - 3 | 251 | 93 | ||
| CN085891.1 | - | 2 - 3 | 318 | 93 | ||
| CR926219.2 | - | 1 - 2 | 229 | 93 | ||
| CX505765.2 | - | 2 - 3 | 350 | 93 | ||
| CX934649.1 | - | 2 - 3 | 310 | 93 | ||
| DT398620.1 | - | 2 - 3 | 316 | 93 | ||
More Information
Set Membership: |
|

hccs
Genomic Map