Assay Details
| Gene Symbol: hccs | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005708.2 | NP_001005708.2 | 6 - 7 | 807 | 81 | |
| NM_001247977.1 | NP_001234906.1 | 6 - 7 | 856 | 81 | ||
| NM_001247978.1 | NP_001234907.1 | 6 - 7 | 810 | 81 | ||
| XM_012960825.2 | XP_012816279.1 | 6 - 7 | 797 | 81 | ||
| GenBank mRNA | BC075286.1 | - | 6 - 7 | 740 | 81 | |
| CR926219.2 | - | 5 - 6 | 718 | 81 | ||
| CX934649.1 | - | 6 - 7 | 799 | 81 | ||
| DT398620.1 | - | 6 - 7 | 805 | 81 | ||
| EL797799.1 | - | 1 - 2 | 405 | 81 | ||
More Information
Set Membership: |
|

hccs
Genomic Map