Assay Details
| Gene Symbol: acadsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005724.1 | NP_001005724.1 | 8 - 9 | 1122 | 68 | |
| XM_012966369.2 | XP_012821823.1 | 8 - 9 | 1185 | 68 | ||
| GenBank mRNA | BC075324.1 | - | 8 - 9 | 1122 | 68 | |
| CR762047.2 | - | 8 - 9 | 982 | 68 | ||
More Information
Set Membership: |
|

acadsb
Genomic Map