Assay Details
| Gene Symbol: pold2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005794.1 | NP_001005794.1 | 2 - 3 | 289 | 71 | |
| GenBank mRNA | BC075341.1 | - | 2 - 3 | 289 | 71 | |
| CR760880.2 | - | 2 - 3 | 252 | 71 | ||
More Information
Set Membership: |
|

pold2
Genomic Map