Assay Details
| Gene Symbol: noxa1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005801.1 | NP_001005801.1 | 9 - 10 | 1014 | 60 | |
| XM_012969757.2 | XP_012825211.1 | 9 - 10 | 1114 | 60 | ||
| XM_018096201.1 | XP_017951690.1 | 10 - 11 | 1183 | 60 | ||
| GenBank mRNA | BC075351.1 | - | 9 - 10 | 1014 | 60 | |
More Information
Set Membership: |
|

noxa1
Genomic Map