Assay Details
Gene Symbol: galk2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005803.1 | NP_001005803.1 | 4 - 5 | 375 | 63 | |
GenBank mRNA | BC075352.1 | - | 4 - 5 | 375 | 63 | |
CT027896.1 | - | 4 - 5 | 450 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
