Assay Details
Gene Symbol: atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005828.2 | NP_001005828.1 | 5 - 6 | 583 | 108 | |
GenBank mRNA | BC075378.1 | - | 5 - 6 | 585 | 108 | |
CT030449.1 | - | 5 - 6 | 587 | 108 | ||
CU075764.1 | - | 5 - 6 | 591 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
