Assay Details
Gene Symbol: atf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005828.2 | NP_001005828.1 | 7 - 8 | 926 | 63 | |
GenBank mRNA | BC075378.1 | - | 7 - 8 | 928 | 63 | |
CT030449.1 | - | 7 - 8 | 930 | 63 | ||
CU075764.1 | - | 7 - 8 | 934 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
