Assay Details
| Gene Symbol: crk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006107.1 | NP_001006107.1 | 1 - 2 | 340 | 78 | |
| XM_012956700.2 | XP_012812154.1 | 1 - 2 | 445 | 78 | ||
| GenBank mRNA | BC074540.1 | - | 1 - 2 | 340 | 78 | |
| CR760069.2 | - | 1 - 2 | 320 | 78 | ||
More Information
Set Membership: |
|

crk
Genomic Map