Assay Details
| Gene Symbol: crk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006107.1 | NP_001006107.1 | 2 - 3 | 881 | 84 | |
| GenBank mRNA | BC074540.1 | - | 2 - 3 | 881 | 84 | |
| CR760069.2 | - | 2 - 3 | 861 | 84 | ||
More Information
Set Membership: |
|

crk
Genomic Map