Assay Details
| Gene Symbol: rab30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006108.1 | NP_001006108.1 | 2 - 3 | 260 | 71 | |
| XM_012956713.2 | XP_012812167.1 | 1 - 2 | 162 | 71 | ||
| XM_012956714.2 | XP_012812168.1 | 2 - 3 | 306 | 71 | ||
| GenBank mRNA | BC074609.1 | - | 2 - 3 | 260 | 71 | |
| CT025302.2 | - | 2 - 3 | 200 | 71 | ||
More Information
Set Membership: |
|

rab30
Genomic Map