Assay Details
Gene Symbol: acta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006708.1 | NP_001006709.1 | 3 - 4 | 495 | 73 | |
GenBank mRNA | BC075427.1 | - | 3 - 4 | 495 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
