Assay Details
Gene Symbol: b4galt3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006718.1 | NP_001006719.1 | 1125 | 58 | ||
GenBank mRNA | BC075452.1 | - | 1125 | 58 | ||
CT027894.1 | - | 1143 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
