Assay Details
Gene Symbol: pnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006719.1 | NP_001006720.1 | 4 - 5 | 628 | 99 | |
XM_012959567.1 | XP_012815021.1 | 3 - 4 | 460 | 99 | ||
GenBank mRNA | BC075454.1 | - | 4 - 5 | 628 | 99 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
