Assay Details
Gene Symbol: apobec2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006721.1 | NP_001006722.1 | 1 - 2 | 197 | 102 | |
GenBank mRNA | BC075459.1 | - | 1 - 2 | 197 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
