Assay Details
Gene Symbol: papss1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006742.1 | NP_001006743.1 | 7 - 8 | 992 | 60 | |
XM_012959851.2 | XP_012815305.1 | 7 - 8 | 1173 | 60 | ||
GenBank mRNA | BC075507.1 | - | 7 - 8 | 992 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
