Assay Details
Gene Symbol: slc9a3r1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006750.1 | NP_001006751.1 | 4 - 5 | 669 | 63 | |
GenBank mRNA | BC075520.1 | - | 4 - 5 | 669 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
