Assay Details
Gene Symbol: pgpep1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006751.1 | NP_001006752.1 | 3 - 4 | 649 | 63 | |
GenBank mRNA | BC075524.1 | - | 3 - 4 | 649 | 63 | |
CT030462.1 | - | 3 - 4 | 267 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
