Assay Details
Gene Symbol: tspan17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006753.1 | NP_001006754.1 | 3 - 4 | 468 | 73 | |
GenBank mRNA | BC075528.1 | - | 3 - 4 | 468 | 73 | |
CT485717.2 | - | 3 - 4 | 476 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
