Assay Details
Gene Symbol: pax6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006762.1 | NP_001006763.1 | 6 - 7 | 901 | 67 | |
XM_012960856.1 | XP_012816310.1 | 4 - 5 | 569 | 67 | ||
XM_012960857.1 | XP_012816311.1 | 3 - 4 | 527 | 67 | ||
XM_012960858.1 | XP_012816312.1 | 4 - 5 | 569 | 67 | ||
XM_012960859.1 | XP_012816313.1 | 3 - 4 | 527 | 67 | ||
XM_018092994.1 | XP_017948483.1 | 7 - 8 | 1142 | 67 | ||
XM_018092995.1 | XP_017948484.1 | 7 - 8 | 946 | 67 | ||
XM_018092996.1 | XP_017948485.1 | 7 - 8 | 981 | 67 | ||
XM_018092997.1 | XP_017948486.1 | 6 - 7 | 1236 | 67 | ||
XM_018092998.1 | XP_017948487.1 | 7 - 8 | 1312 | 67 | ||
XM_018092999.1 | XP_017948488.1 | 8 - 9 | 1184 | 67 | ||
XM_018093000.1 | XP_017948489.1 | 4 - 5 | 594 | 67 | ||
GenBank mRNA | BC075551.1 | - | 6 - 7 | 901 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
