Assay Details
Gene Symbol: ccna2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006767.1 | NP_001006768.1 | 5 - 6 | 1032 | 72 | |
GenBank mRNA | BC075562.1 | - | 5 - 6 | 1032 | 72 | |
CR848597.2 | - | 5 - 6 | 1029 | 72 | ||
CR926264.1 | - | 1 - 2 | 164 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
