Assay Details
Gene Symbol: alg5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006779.1 | NP_001006780.1 | 8 - 9 | 903 | 69 | |
GenBank mRNA | BC075587.1 | - | 8 - 9 | 903 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
