Assay Details
| Gene Symbol: ctdsp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006792.1 | NP_001006793.1 | 6 - 7 | 897 | 66 | |
| XM_012956751.2 | XP_012812205.1 | 7 - 8 | 1359 | 66 | ||
| XM_018091166.1 | XP_017946655.1 | 5 - 6 | 1435 | 66 | ||
| GenBank mRNA | BC076658.1 | - | 6 - 7 | 897 | 66 | |
More Information
Set Membership: |
|

ctdsp2
Genomic Map