Assay Details
Gene Symbol: gstt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006810.1 | NP_001006811.1 | 1 - 2 | 130 | 79 | |
GenBank mRNA | BC076690.1 | - | 1 - 2 | 130 | 79 | |
CR760091.2 | - | 1 - 2 | 137 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
