Assay Details
Gene Symbol: rps9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006812.1 | NP_001006813.1 | 430 | 60 | ||
XM_012967546.1 | XP_012823000.1 | 482 | 60 | |||
GenBank mRNA | BC076696.1 | - | 430 | 60 | ||
CR760722.2 | - | 407 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
