Assay Details
| Gene Symbol: sec22a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006817.1 | NP_001006818.1 | 1 - 2 | 282 | 89 | |
| GenBank mRNA | BC076712.1 | - | 1 - 2 | 282 | 89 | |
More Information
Set Membership: |
|

sec22a
Genomic Map