Assay Details
| Gene Symbol: hal.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006831.2 | NP_001006832.2 | 10 - 11 | 1003 | 91 | |
| NM_001278528.1 | NP_001265457.1 | 11 - 12 | 1022 | 91 | ||
| XM_012958847.2 | XP_012814301.1 | 11 - 12 | 1158 | 91 | ||
| XM_012958848.1 | XP_012814302.1 | 11 - 12 | 971 | 91 | ||
| XM_012958849.2 | XP_012814303.1 | 11 - 12 | 1038 | 91 | ||
| XM_012958850.1 | XP_012814304.1 | 10 - 11 | 850 | 91 | ||
| XM_012958851.1 | XP_012814305.1 | 10 - 11 | 729 | 91 | ||
| XM_018092036.1 | XP_017947525.1 | 11 - 12 | 971 | 91 | ||
| XM_018092037.1 | XP_017947526.1 | 10 - 11 | 933 | 91 | ||
| GenBank mRNA | BC076901.1 | - | 10 - 11 | 1002 | 91 | |
| DN027070.1 | - | 7 - 8 | 527 | 91 | ||
More Information
Set Membership: |
|

hal.2
Genomic Map