Assay Details
| Gene Symbol: hal.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006831.2 | NP_001006832.2 | 2 - 3 | 406 | 110 | |
| NM_001278528.1 | NP_001265457.1 | 3 - 4 | 425 | 110 | ||
| XM_012958847.2 | XP_012814301.1 | 3 - 4 | 561 | 110 | ||
| XM_012958848.1 | XP_012814302.1 | 3 - 4 | 374 | 110 | ||
| XM_012958849.2 | XP_012814303.1 | 3 - 4 | 441 | 110 | ||
| XM_012958851.1 | XP_012814305.1 | 2 - 3 | 132 | 110 | ||
| XM_018092036.1 | XP_017947525.1 | 3 - 4 | 374 | 110 | ||
| XM_018092037.1 | XP_017947526.1 | 3 - 4 | 462 | 110 | ||
| GenBank mRNA | BC076901.1 | - | 2 - 3 | 405 | 110 | |
| CF343491.1 | - | 2 - 3 | 422 | 110 | ||
| DN033227.1 | - | 3 - 4 | 425 | 110 | ||
More Information
Set Membership: |
|

hal.2
Genomic Map