Assay Details
| Gene Symbol: hal.2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006831.2 | NP_001006832.2 | 4 - 5 | 514 | 69 | |
| NM_001278528.1 | NP_001265457.1 | 5 - 6 | 533 | 69 | ||
| XM_012958847.2 | XP_012814301.1 | 5 - 6 | 669 | 69 | ||
| XM_012958848.1 | XP_012814302.1 | 5 - 6 | 482 | 69 | ||
| XM_012958849.2 | XP_012814303.1 | 5 - 6 | 549 | 69 | ||
| XM_012958850.1 | XP_012814304.1 | 4 - 5 | 361 | 69 | ||
| XM_012958851.1 | XP_012814305.1 | 4 - 5 | 240 | 69 | ||
| XM_018092036.1 | XP_017947525.1 | 5 - 6 | 482 | 69 | ||
| XM_018092037.1 | XP_017947526.1 | 5 - 6 | 570 | 69 | ||
| GenBank mRNA | BC076901.1 | - | 4 - 5 | 513 | 69 | |
| CF343491.1 | - | 4 - 5 | 530 | 69 | ||
| DN027070.1 | - | 1 - 2 | 38 | 69 | ||
| DN033227.1 | - | 5 - 6 | 533 | 69 | ||
More Information
Set Membership: |
|

hal.2
Genomic Map