Assay Details
| Gene Symbol: srd5a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006840.1 | NP_001006841.1 | 1 - 2 | 566 | 91 | |
| GenBank mRNA | BC076920.1 | - | 1 - 2 | 566 | 91 | |
| CR761038.2 | - | 1 - 2 | 350 | 91 | ||
More Information
Set Membership: |
|

srd5a1
Genomic Map