Assay Details
Gene Symbol: pgrmc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006841.1 | NP_001006842.1 | 2 - 3 | 525 | 74 | |
XM_012968247.2 | XP_012823701.1 | 2 - 3 | 847 | 74 | ||
XM_012968248.2 | XP_012823702.1 | 2 - 3 | 423 | 74 | ||
XM_012968249.1 | XP_012823703.1 | 2 - 3 | 517 | 74 | ||
XM_012968250.2 | XP_012823704.1 | 2 - 3 | 404 | 74 | ||
XM_012968251.2 | XP_012823705.1 | 2 - 3 | 411 | 74 | ||
XM_012968252.1 | XP_012823706.1 | 2 - 3 | 503 | 74 | ||
GenBank mRNA | BC076926.1 | - | 2 - 3 | 525 | 74 | |
CR762169.2 | - | 342 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
