Assay Details
| Gene Symbol: bcl7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006871.1 | NP_001006872.1 | 3 - 4 | 319 | 73 | |
| GenBank mRNA | BC076987.1 | - | 3 - 4 | 319 | 73 | |
| CR926398.1 | - | 3 - 4 | 260 | 73 | ||
More Information
Set Membership: |
|

bcl7a
Genomic Map