Assay Details
| Gene Symbol: eef1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006876.2 | NP_001006877.1 | 2 - 3 | 247 | 85 | |
| GenBank mRNA | BC077005.1 | - | 2 - 3 | 248 | 85 | |
More Information
Set Membership: |
|

eef1b2
Genomic Map