Assay Details
| Gene Symbol: eif6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006883.1 | NP_001006884.1 | 3 - 4 | 488 | 59 | |
| GenBank mRNA | BC077665.1 | - | 3 - 4 | 488 | 59 | |
| CR760524.2 | - | 3 - 4 | 451 | 59 | ||
| CR848359.2 | - | 4 - 5 | 542 | 59 | ||
More Information
Set Membership: |
|

eif6
Genomic Map