Assay Details
Gene Symbol: mrpl12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006885.1 | NP_001006886.1 | 2 - 3 | 294 | 68 | |
XM_018097712.1 | XP_017953201.1 | 2 - 3 | 379 | 68 | ||
GenBank mRNA | BC077682.1 | - | 2 - 3 | 294 | 68 | |
CR855493.2 | - | 2 - 3 | 287 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
