Assay Details
| Gene Symbol: wrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006891.1 | NP_001006892.1 | 1 - 2 | 139 | 83 | |
| GenBank mRNA | BC077696.1 | - | 1 - 2 | 139 | 83 | |
More Information
Set Membership: |
|

wrb
Genomic Map