Assay Details
| Gene Symbol: tubb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006894.1 | NP_001006895.1 | 3 - 4 | 325 | 85 | |
| XM_012968256.2 | XP_012823710.1 | 3 - 4 | 720 | 85 | ||
| GenBank mRNA | BC074549.1 | - | 3 - 4 | 325 | 85 | |
| CT027951.1 | - | 2 - 3 | 157 | 85 | ||
| CT030380.1 | - | 3 - 4 | 339 | 85 | ||
More Information
Set Membership: |
|

tubb
Genomic Map