Assay Details
| Gene Symbol: gtf2h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006920.1 | NP_001006921.1 | 6 - 7 | 895 | 62 | |
| XM_012954494.2 | XP_012809948.1 | 6 - 7 | 1157 | 62 | ||
| XM_012954495.2 | XP_012809949.1 | 6 - 7 | 1159 | 62 | ||
| GenBank mRNA | BC075586.1 | - | 6 - 7 | 895 | 62 | |
More Information
Set Membership: |
|

gtf2h1
Genomic Map