Assay Details
Gene Symbol: h2afy2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006924.1 | NP_001006925.1 | 7 - 8 | 1100 | 79 | |
GenBank mRNA | BC076893.1 | - | 7 - 8 | 1100 | 79 | |
CR942423.1 | - | 7 - 8 | 986 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
