Assay Details
Gene Symbol: pitx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007499.1 | NP_001007500.1 | 1 - 2 | 372 | 67 | |
GenBank mRNA | BC079936.1 | - | 1 - 2 | 372 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
