Assay Details
Gene Symbol: rcc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007509.1 | NP_001007510.1 | 5 - 6 | 710 | 65 | |
XM_012956782.2 | XP_012812236.1 | 5 - 6 | 735 | 65 | ||
GenBank mRNA | BC079946.1 | - | 5 - 6 | 710 | 65 | |
CT030112.2 | - | 5 - 6 | 682 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
