Assay Details
| Gene Symbol: atp6v1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007511.1 | NP_001007512.1 | 1725 | 62 | ||
| GenBank mRNA | BC079948.1 | - | 1725 | 62 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

atp6v1a| Gene Symbol: atp6v1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007511.1 | NP_001007512.1 | 1725 | 62 | ||
| GenBank mRNA | BC079948.1 | - | 1725 | 62 | ||
Set Membership: |
|