Assay Details
Gene Symbol: naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007512.1 | NP_001007513.1 | 3 - 4 | 204 | 75 | |
XM_012956783.2 | XP_012812237.1 | 7 - 8 | 2532 | 75 | ||
XM_012956784.2 | XP_012812238.1 | 6 - 7 | 2459 | 75 | ||
XM_012956785.2 | XP_012812239.1 | 8 - 9 | 2246 | 75 | ||
XM_012956786.2 | XP_012812240.1 | 6 - 7 | 829 | 75 | ||
XM_012956787.2 | XP_012812241.1 | 5 - 6 | 760 | 75 | ||
XM_012956788.2 | XP_012812242.1 | 5 - 6 | 673 | 75 | ||
XM_012956789.1 | XP_012812243.1 | 4 - 5 | 604 | 75 | ||
XM_012956790.1 | XP_012812244.1 | 4 - 5 | 421 | 75 | ||
GenBank mRNA | BC079953.1 | - | 3 - 4 | 204 | 75 | |
BC167646.1 | - | 4 - 5 | 1471 | 75 | ||
CT025482.2 | - | 3 - 4 | 169 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
