Assay Details
Gene Symbol: dek | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007865.1 | NP_001007866.1 | 8 - 9 | 1089 | 72 | |
GenBank mRNA | BC079968.1 | - | 8 - 9 | 1089 | 72 | |
CR760931.2 | - | 8 - 9 | 1060 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
