Assay Details
| Gene Symbol: cbr4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007872.1 | NP_001007873.1 | 4 - 5 | 435 | 80 | |
| XM_012960249.2 | XP_012815703.1 | 4 - 5 | 513 | 80 | ||
| XM_012960250.2 | XP_012815704.1 | 4 - 5 | 662 | 80 | ||
| XM_012960257.2 | XP_012815711.1 | 4 - 5 | 598 | 80 | ||
| GenBank mRNA | BC080139.1 | - | 4 - 5 | 435 | 80 | |
More Information
Set Membership: |
|

cbr4
Genomic Map