Assay Details
| Gene Symbol: rab25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001007902.1 | NP_001007903.1 | 2 - 3 | 425 | 65 | |
| XM_018096226.1 | XP_017951715.1 | 2 - 3 | 509 | 65 | ||
| GenBank mRNA | BC080339.1 | - | 2 - 3 | 425 | 65 | |
| CR848299.2 | - | 2 - 3 | 414 | 65 | ||
More Information
Set Membership: |
|

rab25
Genomic Map